CH4238556855 ChemBridge Corporation

3-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

  • MDL number MFCD00023096
  • Cas Number : 3339-73-9
  • Molecular Weight : 219.1935

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005053

Safety Information

MSDS https://www.hit2lead.com/sds/3005053_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart