CH4238556908 ChemBridge Corporation

(2E)-3-(1-ethyl-1H-pyrazol-4-yl)acrylic acid

  • MDL number MFCD00462200
  • Cas Number : 1613051-24-3
  • Molecular Weight : 166.1772

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005129

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3005129
Availability details for
Quantity
  • Availability
  • Add to Cart