CH4238556940 ChemBridge Corporation

5-(2-methoxyphenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD07377106
  • Cas Number : 668971-16-2
  • Molecular Weight : 219.1935

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005171

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3005171
Availability details for
Quantity
  • Availability
  • Add to Cart