CH4238556961 ChemBridge Corporation

3-(4-fluorophenyl)-5-methyl-4,5-dihydroisoxazole-5-carboxylic acid

  • MDL number MFCD06753283
  • Cas Number : 878426-96-1
  • Molecular Weight : 223.2004

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005200

Safety Information

MSDS https://www.hit2lead.com/sds/3005200_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart