CH4238556983 ChemBridge Corporation

(2E)-3-(1,3-dimethyl-1H-pyrazol-4-yl)acrylic acid

  • MDL number MFCD02055619
  • Cas Number : 1613051-25-4
  • Molecular Weight : 166.1772

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005229

Safety Information

MSDS https://www.hit2lead.com/sds/3005229_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart