CH4238556987 ChemBridge Corporation

7-(2-thienyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD04049989
  • Cas Number : 676247-93-1
  • Molecular Weight : 245.2572

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005233

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3005233
Availability details for
Quantity
  • Availability
  • Add to Cart