CH4238557077 ChemBridge Corporation

3-chloro-5-phenyl-7-(trifluoromethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD01922903
  • Cas Number : 312699-22-2
  • Molecular Weight : 345.7042

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005365

Safety Information

MSDS https://www.hit2lead.com/sds/3005365_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart