CH4238557186 ChemBridge Corporation

3-(1,3-dimethyl-1H-pyrazol-4-yl)-5-methylisoxazole-4-carboxylic acid

  • MDL number MFCD12027175
  • Cas Number : 1171972-67-0
  • Molecular Weight : 221.2126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020098

Safety Information

MSDS https://www.hit2lead.com/sds/3020098_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart