CH4238557197 ChemBridge Corporation

1-(pyrrolidin-1-ylsulfonyl)piperidine-3-carboxylic acid

  • MDL number MFCD11171650
  • Cas Number : 1042640-05-0
  • Molecular Weight : 262.3259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020116

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3020116
Availability details for
Quantity
  • Availability
  • Add to Cart