CH4238557248 ChemBridge Corporation

4-(2-furyl)-N-methylbutan-2-amine

  • MDL number MFCD00520659
  • Cas Number : 1593-37-9
  • Molecular Weight : 153.2215

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020258

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3020258
Availability details for
Quantity
  • Availability
  • Add to Cart