CH4238557305 ChemBridge Corporation

1-(2-piperidin-2-ylethyl)piperidine

  • MDL number MFCD00083062
  • Cas Number : 14759-07-0
  • Molecular Weight : 196.3324

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020321

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3020321
Availability details for
Quantity
  • Availability
  • Add to Cart