CH4238557328 ChemBridge Corporation

5-(dimethylamino)thiophene-2-carbaldehyde

  • MDL number MFCD00708772
  • Cas Number : 24372-46-1
  • Molecular Weight : 155.2175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020346

Safety Information

MSDS https://www.hit2lead.com/sds/3020346_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart