CH4238557358 ChemBridge Corporation

1-(piperazin-1-ylacetyl)azepane

  • MDL number MFCD07685910
  • Cas Number : 39890-47-6
  • Molecular Weight : 225.3305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020376

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3020376
Availability details for
Quantity
  • Availability
  • Add to Cart