CH4238557439 ChemBridge Corporation

4,6-dimethyl-5-morpholin-4-ylpyrimidin-2-amine

  • MDL number MFCD12027400
  • Cas Number : 1119451-57-8
  • Molecular Weight : 208.2602

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3020467

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3020467
Availability details for
Quantity
  • Availability
  • Add to Cart