CH4238557467 ChemBridge Corporation

(3-oxo-2-piperazinyl)acetic acid

  • MDL number MFCD04226363
  • Cas Number : 405214-33-7
  • Molecular Weight : 158.1552

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004068

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004068
Availability details for
Quantity
  • Availability
  • Add to Cart