CH4238557481 ChemBridge Corporation

3-(chloromethyl)-5-ethyl-1,2,4-oxadiazole

  • MDL number MFCD11845661
  • Cas Number : 83227-01-4
  • Molecular Weight : 146.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(chloromethyl)-5-ethyl-1,2,4-oxadiazole
Inchi Code 1S/C5H7ClN2O/c1-2-5-7-4(3-6)8-9-5/h2-3H2,1H3
Inchi Key PGYXLMWYFOXGGN-UHFFFAOYSA-N

Description

Purity 95%
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4013190

Safety Information

MSDS https://www.hit2lead.com/sds/4013190_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart