CH4238557519 ChemBridge Corporation

(1-cyclohexyl-3-pyrrolidinyl)methanol

  • MDL number MFCD10003885
  • Cas Number : 100049-71-6
  • Molecular Weight : 183.2905

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026408

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4026408
Availability details for
Quantity
  • Availability
  • Add to Cart