CH4238557532 ChemBridge Corporation

1-methyl-1H-indole-6-carboxylic acid

  • MDL number MFCD08690250
  • Cas Number : 202745-73-1
  • Molecular Weight : 175.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029335

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029335
Availability details for
Quantity
  • Availability
  • Add to Cart