CH4238557535 ChemBridge Corporation

N-(methylsulfonyl)alanine

  • MDL number MFCD07403634
  • Cas Number : 879123-60-1
  • Molecular Weight : 167.1836

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030456

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4030456
Availability details for
Quantity
  • Availability
  • Add to Cart