CH4238557541 ChemBridge Corporation

3-(2-aminophenyl)-N-methylpropanamide

  • MDL number MFCD10022153
  • Cas Number : 1018506-37-0
  • Molecular Weight : 178.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032084

Safety Information

MSDS https://www.hit2lead.com/sds/4032084_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart