CH4238557581 ChemBridge Corporation

3-aminothieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD03783349
  • Cas Number : 58327-75-6
  • Molecular Weight : 194.2104

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033010

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033010
Availability details for
Quantity
  • Availability
  • Add to Cart