CH4238557600 ChemBridge Corporation

2,5-dichloro-1,3-benzoxazole

  • MDL number MFCD09264312
  • Cas Number : 3621-81-6
  • Molecular Weight : 188.0108

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3003003

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3003003
Availability details for
Quantity
  • Availability
  • Add to Cart