CH4238557648 ChemBridge Corporation

({1-[(5-methyl-2-furyl)methyl]-4-piperidinyl}methyl)amine

  • MDL number MFCD12195858
  • Cas Number : 1185320-32-4
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034628

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034628
Availability details for
Quantity
  • Availability
  • Add to Cart