CH4238557654 ChemBridge Corporation

3-[(dimethylamino)carbonyl]-4-methoxybenzenesulfonyl chloride

  • MDL number MFCD12827497
  • Cas Number : 1114594-34-1
  • Molecular Weight : 277.7246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034831

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4034831_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart