CH4238557656 ChemBridge Corporation

2-(4-ethyl-3,5-dimethyl-1H-pyrazol-1-yl)ethanamine

  • MDL number MFCD12030901
  • Cas Number : 562815-62-7
  • Molecular Weight : 167.2514

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034837

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034837
Availability details for
Quantity
  • Availability
  • Add to Cart