CH4238557676 ChemBridge Corporation

1,7-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrido[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD08548408
  • Cas Number : 110181-69-6
  • Molecular Weight : 235.1962

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035029

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035029
Availability details for
Quantity
  • Availability
  • Add to Cart