CH4238557685 ChemBridge Corporation

2-isopropyl-4-methyl-5-pyrimidinecarboxylic acid

  • MDL number MFCD09750210
  • Cas Number : 954233-00-2
  • Molecular Weight : 180.2038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035107

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035107
Availability details for
Quantity
  • Availability
  • Add to Cart