CH4238557701 ChemBridge Corporation

5-(1,5-dimethyl-1H-pyrazol-4-yl)-3-isoxazolecarboxylic acid

  • MDL number MFCD04969981
  • Cas Number : 957312-71-9
  • Molecular Weight : 207.1861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035263

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035263
Availability details for
Quantity
  • Availability
  • Add to Cart