CH4238557727 ChemBridge Corporation

6-bromo-3-pyridazinol

  • MDL number MFCD11223475
  • Cas Number : 51355-94-3
  • Molecular Weight : 174.9834

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035405

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035405
Availability details for
Quantity
  • Availability
  • Add to Cart