CH4238557735 ChemBridge Corporation

4-(acetylamino)-1-naphthalenesulfonyl chloride

  • MDL number MFCD02169752
  • Cas Number : 5690-20-0
  • Molecular Weight : 283.7307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035512

Safety Information

signalword Danger
MSDS https://www.hit2lead.com/building-blocks/4035512
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart