CH4238557741 ChemBridge Corporation

1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02090853
  • Cas Number : 128694-63-3
  • Molecular Weight : 194.1113

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035600

Safety Information

MSDS https://www.hit2lead.com/sds/4035600_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart