CH4238557752 ChemBridge Corporation

7-hydroxy-3-quinolinecarbaldehyde

  • MDL number MFCD10696340
  • Cas Number : 363135-54-0
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035676

Safety Information

MSDS https://www.hit2lead.com/sds/4035676_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart