CH4238557765 ChemBridge Corporation

6-chloro-N,N-dimethyl-2-pyrazinamine

  • MDL number MFCD12024889
  • Cas Number : 61655-72-9
  • Molecular Weight : 157.6008

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035795

Safety Information

MSDS https://www.hit2lead.com/sds/4035795_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart