CH4238557767 ChemBridge Corporation

1,5-dimethyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD04968668
  • Cas Number : 1005613-94-4
  • Molecular Weight : 194.6393

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035817

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4035817_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart