CH4238557769 ChemBridge Corporation

1-ethyl-3-methyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD04968671
  • Cas Number : 1005627-55-3
  • Molecular Weight : 208.6659

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035828

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4035828_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart