CH4238557822 ChemBridge Corporation

1-(4-pyridinyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD07186449
  • Cas Number : 842972-67-2
  • Molecular Weight : 172.1833

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4302044

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4302044
Availability details for
Quantity
  • Availability
  • Add to Cart