CH4244052228 ChemBridge Corporation

6-bromo-N-methyl-3-pyridazinamine

  • MDL number MFCD13188585
  • Cas Number : 1208657-07-1
  • Molecular Weight : 188.0252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035399

Safety Information

MSDS https://www.hit2lead.com/sds/4035399_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart