CH4244052252 ChemBridge Corporation

2-chloro-1,3-thiazole-4-carboxamide

  • MDL number MFCD13188626
  • Cas Number : 928256-35-3
  • Molecular Weight : 162.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036343

Safety Information

MSDS https://www.hit2lead.com/sds/4036343_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart