CH4244052258 ChemBridge Corporation

3-(5-isoxazolyl)propanoic acid

  • MDL number MFCD07788406
  • Cas Number : 98140-73-9
  • Molecular Weight : 141.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4018161

Safety Information

MSDS https://www.hit2lead.com/sds/4018161_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart