CH4244052264 ChemBridge Corporation

4-chloro-6-(4-morpholinyl)-2-pyrimidinamine

  • MDL number MFCD00664697
  • Cas Number : 339016-18-1
  • Molecular Weight : 214.6521

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4020051

Safety Information

msds https://www.hit2lead.com/building-blocks/4020051
Availability details for
Quantity
  • Availability
  • Add to Cart