CH4244052318 ChemBridge Corporation

1-methyltetrahydro-2(1H)-pyrimidinone

  • MDL number MFCD00196814
  • Cas Number : 10166-54-8
  • Molecular Weight : 114.1457

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035075

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035075
Availability details for
Quantity
  • Availability
  • Add to Cart