CH4244052330 ChemBridge Corporation

5-methyl-1,3-benzoxazol-2-amine

  • MDL number MFCD01664145
  • Cas Number : 64037-15-6
  • Molecular Weight : 148.1619

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4007276

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4007276
Availability details for
Quantity
  • Availability
  • Add to Cart