CH4244052337 ChemBridge Corporation

2-methyl-4,5,6,7-tetrahydro-2H-indazole-3-carboxylic acid

  • MDL number MFCD04092072
  • Cas Number : 32287-00-6
  • Molecular Weight : 180.2038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009746

Safety Information

MSDS https://www.hit2lead.com/sds/4009746_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart