CH4244052339 ChemBridge Corporation

tert-butyl [(6-chloro-3-pyridinyl)methyl]carbamate

  • MDL number MFCD11974332
  • Cas Number : 285119-72-4
  • Molecular Weight : 242.702

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4035250

Safety Information

msds https://www.hit2lead.com/building-blocks/4035250
Availability details for
Quantity
  • Availability
  • Add to Cart