CH4244052343 ChemBridge Corporation

4-chloro-1H-indole-2,3-dione

  • MDL number MFCD00129178
  • Molecular Weight : 181.5759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035272

Safety Information

MSDS https://www.hit2lead.com/sds/4035272_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart