CH4244052378 ChemBridge Corporation

2-(1H-1,2,4-triazol-3-yl)pyridine

  • MDL number MFCD13188589
  • Cas Number : 23195-62-2
  • Molecular Weight : 146.1493

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035532

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035532
Availability details for
Quantity
  • Availability
  • Add to Cart