CH4244052404 ChemBridge Corporation

3-butynoic acid

  • MDL number MFCD02258494
  • Cas Number : 2345-51-9
  • Molecular Weight : 84.0734

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035749

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035749
Availability details for
Quantity
  • Availability
  • Add to Cart