CH4244052428 ChemBridge Corporation

2-cyclopropyl-5-pyrimidinecarboxylic acid

  • MDL number MFCD09054852
  • Cas Number : 648423-79-4
  • Molecular Weight : 164.1613

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035935

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4035935
Availability details for
Quantity
  • Availability
  • Add to Cart