CH4244052443 ChemBridge Corporation

7-quinolinecarbonitrile

  • MDL number MFCD12028057
  • Cas Number : 67360-38-7
  • Molecular Weight : 154.168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036036

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4036036_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart