CH4244052447 ChemBridge Corporation

6-chloro-N~4~-isopropyl-2,4-pyrimidinediamine

  • MDL number MFCD13188621
  • Cas Number : 30182-24-2
  • Molecular Weight : 186.642

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4036075

Safety Information

MSDS https://www.hit2lead.com/sds/4036075_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart